Accessories for Level Control Signal Boxes
Waste disposal set 7: (1) funnel with lid MARCO, S60/61, (1) canister, S60/61, 10 L, (1) collecting tray with base insert
Catalog # 76948-914
Supplier: SCAT AMERICAS, INC.
Supplier #: 307315
Jump to:
Product Details & Documents
Useful accessories, extensions, and alternative products for the SCAT level control signal boxes.
- Cable for relay contacts
- Disc sensors
- Dual lock
- Signal cables
- Power supplies
- Velcro strips
Ensure SCAT level control signal boxes are perfectly equipped.
Documents
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Specifications
- Description:Waste disposal set 7: (1) funnel with lid MARCO, S60/61, (1) canister, S60/61, 10 L, (1) collecting tray with base insert
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



