Orders LinkContactUsLinkComponent
Control Slides IHC CD 23
Control Slides IHC CD 23
Control slides IHC CD 23
Catalog # 76945-676
Supplier:   Mopec
Supplier #:  SC085-100
Control Slides IHC CD 23
Catalog # 76945-676
Supplier:  Mopec
Control Slides IHC CD 23
Control Slides IHC CD 23
Control slides IHC CD 23
Price
$1,640.22
Log in for availability
Case of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
All control slides are on Mopec IHC enhanced hydrophilic adhesive slides using formalin fixed paraffin embedded (FFPE) human tissue.
  • Product specific reactivity is guaranteed for one year from date of receipt
  • Revalidate after one year to verify continued reactivity
  • All controls are cut in serial sections at 4 µm
  • Standard tissue placement is mid-high on slide to facilitate placement of patient and control tissue on the same slide
  • All slides are air dried
Product specific reactivity is guaranteed for one year from date of receipt when stored properly at 60 to 80 °F in a humidity controlled and light-deprived environment.
Packaging: Case of 25 contains 24 unstained slides and 1 stained slide. Case of 100 contains 98 unstained slides and 2 stained slides.
Specifications
  • Description:
    Control slides IHC CD 23