Orders LinkContactUsLinkComponent
Amplitude™ Lambda™ Wipes
Amplitude™ Lambda™ Wipes
Amplitude™ Lambda™ wipes, flat stacked, Cellulose/polyester, 17.8×17.8 cm (7×7")
Catalog # 76945-154
Amplitude™ Lambda™ Wipes
Catalog # 76945-154
Supplier:  
Amplitude™ Lambda™ Wipes
Amplitude™ Lambda™ Wipes
Amplitude™ Lambda™ wipes, flat stacked, Cellulose/polyester, 17.8×17.8 cm (7×7")
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
WIPE NW LAMBDA* CELL/POLY 9X9IN CS3600 Catalog #10797-028
Product 10797-028 is similar to 76945-154 but manufacturer and specifications may vary.
Product Details & Documents
Amplitude™ Lambda™ Wipes are a hydroentangled blend of 58% cellulose and 42% polyester. The Lambda™ wipe is highly sorbent, has excellent durability, and good cleanliness.
  • Hydroentangled fabric provides low levels of particles and extractables
  • Excellent for wiping and absorbing spills
  • Strong and durable both wet and dry
  • Good cost effective, general-purpose wipe
An excellent all-purpose wipe with outstanding sorbency, it is ideal for general-purpose cleaning and spill pick-up in biotech, pharmaceutical, and medical device industries.
Recommended for ISO Class 6 – 8 environments.
Documents
Specifications
  • Description:
    Amplitude™ Lambda™ wipes, flat stacked
  • Material:
    Cellulose/polyester
  • W×L:
    17.8×17.8 cm (7×7")