Amplitude™ Lambda™ Wipes
Amplitude™ Lambda™ wipes, flat stacked, Cellulose/polyester, 30.5×30.5 cm (12×12")
Catalog # 76945-152
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
WIPE NONWVN PES/CELLULOSE 12X12IN CS2700 Catalog #76470-816
Product 76470-816 is similar to 76945-152 but manufacturer and specifications may vary.
Product Details & Documents
Amplitude™ Lambda™ Wipes are a hydroentangled blend of 58% cellulose and 42% polyester. The Lambda™ wipe is highly sorbent, has excellent durability, and good cleanliness.
- Hydroentangled fabric provides low levels of particles and extractables
- Excellent for wiping and absorbing spills
- Strong and durable both wet and dry
- Good cost effective, general-purpose wipe
An excellent all-purpose wipe with outstanding sorbency, it is ideal for general-purpose cleaning and spill pick-up in biotech, pharmaceutical, and medical device industries.
Recommended for ISO Class 6 – 8 environments.
Documents
Certificates
Search by Lot NumberSpecifications
- Description:Amplitude™ Lambda™ wipes, flat stacked
- Material:Cellulose/polyester
- W×L:30.5×30.5 cm (12×12")