Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
SteraMist BIT Solution

SteraMist BIT Solution

SteraMist BIT Solution for use with all SteraMist Systems, --, 55 gal., 550 lbs.
Catalog # 76919-440
Supplier #:  BIT-40055GL
SteraMist BIT Solution
Catalog # 76919-440
SteraMist BIT Solution
SteraMist BIT Solution

SteraMist BIT Solution for use with all SteraMist Systems, --, 55 gal., 550 lbs.

Price
$9,862.35
Log in for availability
Each(1/Itm/Bx)
Product Details & Documents
  • All SteraMist products exclusively use low-percentage 7.8% Hydrogen Peroxide as a sole active ingredient
  • Each SteraMist product utilizes the same BIT™ Solution blend for a wide array of applications
  • BIT Solution kills within seconds of contact
  • Non-corrosive to enure superior surface compatibility
  • No toxic byproducts
  • No mixing or dilution
  • No wiping or rinsing
Specifications
  • Description:
    SteraMist BIT Solution for use with all SteraMist Systems
  • Size:
    --
  • Volume:
    55 gal.
  • Weight:
    550 lbs.
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.