RSA-Pro X™ Micro-Sample Management Systems and Inserts
Micro-Sample Management System. Complete set, includes 96 individual, 350 µl glass inserts with RSA-Pro X™ surfaces in a rack and base plus a matching solvent resistant, polymeric plug style, sealing mat
Catalog # 76881-754
Supplier: MicroSolv Technology Corp
Supplier #: 95296-350-1PR
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Catalog # 76881-754
Supplier: MicroSolv Technology Corp
Jump to:
Product Details & Documents
Specifications
- Description:Micro-Sample Management System. Complete set, includes 96 individual, 350 µl glass inserts with RSA-Pro X™ surfaces in a rack and base plus a matching solvent resistant, polymeric plug style, sealing mat