Orders LinkContactUsLinkComponent
RSA-Pro X™ Micro-Sample Management Systems and Inserts
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Micro-Sample Management System. Complete set, includes 96 individual, 500 µl glass Inserts with RSA-Pro X™ surfaces in a rack and base plus a matching pre-slit, solvent resistant, polymeric plug style, sealing mat
Catalog # 76881-770
Supplier #:  95296-500-3PR
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Catalog # 76881-770
RSA-Pro X™ Micro-Sample Management Systems and Inserts
Micro-Sample Management System. Complete set, includes 96 individual, 500 µl glass Inserts with RSA-Pro X™ surfaces in a rack and base plus a matching pre-slit, solvent resistant, polymeric plug style, sealing mat
Price
$136.89
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Specifications
  • Description:
    Micro-Sample Management System. Complete set, includes 96 individual, 500 µl glass Inserts with RSA-Pro X™ surfaces in a rack and base plus a matching pre-slit, solvent resistant, polymeric plug style, sealing mat