4-Nitrobenzoic acid 98%
Glass bottle, 5 g
Catalog # 76816-942
Supplier #: A315136-5G
Jump to:
Discontinued
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Caution: for Research Use Only
- CAS Number:62-23-7
- Formula:C₇H₅NO₄
- MDL Number:MFCD00007352
- Molecular Weight:167.12 g/mol
- Boiling Pt:359.1 °C (760 mm Hg)
- Melting Pt:237…240 °C
- Flash Pt:237 °C
- Storage Temperature:Ambient
Specifications
Specifications
- Pack type:Glass bottle
- Size:5 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



