Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SOLVENT DISPENSER IC ESD SAFE 8OZ CS50
Catalog # 76808-238
Supplier:
R & R Lotion
Expand image
SOLVENT DISPENSER IC ESD SAFE 8OZ CS50
Catalog # 76808-238
Supplier:
R & R Lotion
Supplier Number:
SD-8-ESD