Human Recombinant PPIL2 (from E. coli), Unconjugated
200 μg
76795-288
: Biortus
: BP08543-00A-2B
: Frozen liquid on dry ice.
- Size:200 μg
- Cat. No.:76795-288
- Shipping Temperature:Frozen liquid with dry ice
- Protein/Peptide Type:Recombinant
- Gene ID:UniProt Q13356
- Formulation:50 mM Tris-HCl (pH 7.5), 1 mM DTT, 200 mM NaCl, 5% glycerol
- Purity:85%
- Protein/Peptide Name:PPIL2
- Conjugation:Unconjugated
- Sequence:G2-W520
- Species:Human
- Source:E. coli



