Orders LinkContactUsLinkComponent
D-(-)-Sorbitol 95%
Glass bottle, 25 g
  76722-688
 :   Ambeed
 :  A256763-25G
D-(-)-Sorbitol 95%
  76722-688
 :  Ambeed
 :  50-70-4
D-(-)-Sorbitol 95%
Glass bottle, 25 g
 :  50-70-4
$12.00
Each (25g)
 : for Research Use Only
 :  (2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol, (Sorbitol), D(-)-Glucitol, D-Sorbitol, Sorbitol
  •  :50-70-4
  •  :HOH₂C(CH(OH))₄CH₂OH
  •  :MFCD00004708
  •  :182.17 g/mol
  • Pack type:
    Glass bottle
  • Size:
    25 g