Orders LinkContactUsLinkComponent
α-Cyclodextrin 97%
Glass bottle, 100 g
  76689-954
 :   Ambeed
 :  A164223-100G
α-Cyclodextrin 97%
  76689-954
 :  Ambeed
 :  10016-20-3
α-Cyclodextrin 97%
α-Cyclodextrin 97%
Glass bottle, 100 g
 :  10016-20-3
$69.00
Each (100g)
 : for Research Use Only
  •  :10016-20-3
  •  :C₃₆H₆₀O₃₀
  •  :MFCD00078207
  •  :972.84 g/mol
  • Pack type:
    Glass bottle
  • Size:
    100 g