$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
MANIFOLD WASTE MULTI-INLET W/4 STANDARD
MSPP-MW0406
:
ANALYTICAL SALES & SERVICE MS
MANIFOLD WASTE MULTI-INLET W/4 STANDARD
MSPP-MW0406
:
ANALYTICAL SALES & SERVICE MS
:
MW0406