Orders LinkContactUsLinkComponent
Vector PCX Derivatization Instruments
Vector PCX Derivatization Instruments
Vector, single pump, OPA flow restrictor, 0.15 ml, 120 V (primary amino acids w/OPA, aminoglycoside antibiotics, biogenic amines, sulfonamide artificial sweetener)
Catalog # MSPP-1154-4011
Marketsource
Supplier:   Pickering Labs
Supplier #:  1154-4011
Vector PCX Derivatization Instruments
Catalog # MSPP-1154-4011
Supplier:  Pickering Labs
Marketsource
Vector PCX Derivatization Instruments
Vector, single pump, OPA flow restrictor, 0.15 ml, 120 V (primary amino acids w/OPA, aminoglycoside antibiotics, biogenic amines, sulfonamide artificial sweetener)
Price
$29,661.54
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Vector PCX provides the selectivity and sensitivity required for most standard post-column applications while being reliable and easy to use. Since Vector PCX does not have a column oven it is important to use the HPLC column oven to ensure stable column temperature and prevent retention time drifts and separation problems.
  • Independently adjustable, low-pulsation inert pumps
  • PEEK liquid manifolds, Integrated gas manifold
  • Easy access and monitoring of fluidics
  • Simple control interface (no computer required)
  • Reduced bench space profile
  • Sapphire pistons with automatic piston wash
Dimensions: 43×21.6×41.2 cm (17×8.75×16 inches). Weight: 12.5 kg (27.6 lbs) – Duplex. Electrical: 100 to 120 V, 50/60 Hz 1.7 A, 200 W or 200 to 240 V, 50/60 Hz, 0.8 A, 200 W. Mains voltage ±10% of nominal. Installation (over voltage) category II, pollution degree 2. Environmental: altitude up to 6500 ft. Ambient temperature 15 to 25 ºC. Relative humidity up to 80% at 31 ºC. Adjustable from 0.05 to 2.00 ml/minute against back-pressures of up to 2000 psi.
One liter capacity (2 L reservoirs available). Maintained under inert gas pressure to inhibit oxidation of oxygensensitive reagents. Valve built into reservoir cap permits sparging during reagent preparation. Reagent reservoirs fitted with 3.1 mm (¹/₈") Ext.Ø, oxygen-impermeable Air Barrier tubing for oxygen-sensitive reagents and/or with 3.1 mm (¹/₈") Ext.Ø FEP tubing.
Specifications
  • Description:
    Vector, single pump, OPA flow restrictor, 0.15 ml, 120 V (primary amino acids w/OPA, aminoglycoside antibiotics, biogenic amines, sulfonamide artificial sweetener)