Orders LinkContactUsLinkComponent
Osmometer Calibration Standards
Osmometer Calibration Standards
Osmometer calibration standard 1000 mOsm/kg, 10×1 ml ampule
Catalog # 76670-512
Osmometer Calibration Standards
Catalog # 76670-512
Supplier:  
CAS Number:  
Osmometer Calibration Standards
Osmometer Calibration Standards
Osmometer calibration standard 1000 mOsm/kg, 10×1 ml ampule
CAS Number:  
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Omolality calibration standards for clinical and research osmometers.
  • Includes Certificate of Analysis
  • For IVD and research use
  • Compatible with all Osmometers
  • 10×1 ml ampules (ideal volume for unit calibration)
  • Sturdy packaging
  • Two year shelf life
Four E's is a global manufacturer of clinical and research equipment and reagents. These IVD grade Osmometer Calibration Standards are for calibrating Osmometers, they are suitable for use with the Four E's Osmometer or any other Osmometers.
Certifications: CoA and Accuracy statement.
Packaging: Ampules are packaged in a cardboard box.
Caution: Do not shake prior to use, only swirl.
Documents
Specifications
Specifications
  • Description:
    Osmometer calibration standard 1000 mOsm/kg
  • Size:
    10×1 ml ampule