Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Osmometer Calibration Standards

Osmometer Calibration Standards

Osmometer calibration standard 300 mOsm/kg, 10×1 ml ampule
Catalog # 76670-498
Supplier #:  3MA0034ES
Osmometer Calibration Standards
Catalog # 76670-498
CAS Number:  
Osmometer Calibration Standards
Osmometer Calibration Standards

Osmometer calibration standard 300 mOsm/kg, 10×1 ml ampule

CAS Number:  
Price
$198.92
Log in for availability
Box of 10 (1ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Omolality calibration standards for clinical and research osmometers.

  • Includes Certificate of Analysis
  • For IVD and research use
  • Compatible with all Osmometers
  • 10×1 ml ampules (ideal volume for unit calibration)
  • Sturdy packaging
  • Two year shelf life

Four E's is a global manufacturer of clinical and research equipment and reagents. These IVD grade Osmometer Calibration Standards are for calibrating Osmometers, they are suitable for use with the Four E's Osmometer or any other Osmometers.

Certifications: CoA and Accuracy statement.

Packaging: Ampules are packaged in a cardboard box.

Caution: Do not shake prior to use, only swirl.

Documents

Specifications

Specifications

  • Description:
    Osmometer calibration standard 300 mOsm/kg
  • Size:
    10×1 ml ampule

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.