Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Osmometer Calibration Standards

Osmometer calibration standard 1500 mOsm/kg, 10×1 ml ampule
  76670-492
 :  3LA1514ES
Osmometer Calibration Standards
  76670-492
 :  
Osmometer Calibration Standards
Osmometer Calibration Standards

Osmometer calibration standard 1500 mOsm/kg, 10×1 ml ampule

 :  
 
$198.92
   
Box of 10 (1ml)
 
   
 

Omolality calibration standards for clinical and research osmometers.

  • Includes Certificate of Analysis
  • For IVD and research use
  • Compatible with all Osmometers
  • 10×1 ml ampules (ideal volume for unit calibration)
  • Sturdy packaging
  • Two year shelf life

Four E's is a global manufacturer of clinical and research equipment and reagents. These IVD grade Osmometer Calibration Standards are for calibrating Osmometers, they are suitable for use with the Four E's Osmometer or any other Osmometers.

 : CoA and Accuracy statement.

 : Ampules are packaged in a cardboard box.

 : Do not shake prior to use, only swirl.

 

 

 

  • Description:
    Osmometer calibration standard 1500 mOsm/kg
  • Size:
    10×1 ml ampule

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.