Orders LinkContactUsLinkComponent
Osmometer Calibration Standards
Osmometer Calibration Standards
Osmometer calibration standard, two level: 300/800 mOsm/kg, 10×1 ml ampule
  76670-488
Osmometer Calibration Standards
  76670-488
 :  
 :  
Osmometer Calibration Standards
Osmometer Calibration Standards
Osmometer calibration standard, two level: 300/800 mOsm/kg, 10×1 ml ampule
 :  
Omolality calibration standards for clinical and research osmometers.
  • Includes Certificate of Analysis
  • For IVD and research use
  • Compatible with all Osmometers
  • 10×1 ml ampules (ideal volume for unit calibration)
  • Sturdy packaging
  • Two year shelf life
Four E's is a global manufacturer of clinical and research equipment and reagents. These IVD grade Osmometer Calibration Standards are for calibrating Osmometers, they are suitable for use with the Four E's Osmometer or any other Osmometers.
Certifications: CoA and Accuracy statement.
Packaging: Ampules are packaged in a cardboard box.
Caution: Do not shake prior to use, only swirl.
Documents
  • Description:
    Osmometer calibration standard, two level: 300/800 mOsm/kg
  • Size:
    10×1 ml ampule