COLUMN HPLC INERTSIL 3UM 150 4.6 MM
MSPP-502004045
Frequently Bought Together
VWR® Crimp Caps for 11 mm Crimp Top Vials
: VWR International
Closures for 2 ml Screw Top Vials, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
VWR® 9 mm Screw-Thread Vials, Kits, Caps, and Inserts, Wide Opening
: VWR International
Sodium chloride ≥99.0% ACS analytical reagent, VWR Chemicals BDH®
: VWR International
Screw Top Vials and Caps, 2 ml, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Disposable Plain Centrifuge Tubes, Glass, Kimble Chase
: DWK Life Sciences (KIMBLE)
VWR® Screw Caps for 9 mm Screw-Thread Vials, Wide Opening
: VWR International
GC Autosampler Syringes, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Gas Clean Purification System, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
Fused Silica Tubing Cutters for GC, Agilent Technologies
: AGILENT TECHNOLOGIES, INC (CSD)
CrossLab Graphite/Vespel Ferrules for GC
: AGILENT TECHNOLOGIES, INC (CSD)
VWR® 11 mm Snap Ring Vials, Kits, Caps, and Inserts, Wide Opening
: VWR International
Ammonium formate 99%
: Thermo Scientific Chemicals
SterEM™ Tryptic Soy Agar (TSA), USP, Irradiated
: Hardy Diagnostics