Orders LinkContactUsLinkComponent
Bioanalyzer High Sensitivity DNA Analysis
Bioanalyzer High Sensitivity DNA Analysis
Agilent High sensitivity DNA kit, For the separation, sizing and quantification of dsDNA samples of limited abundance and sized from 50 - 7000 bp, the kit includes 10 microfluidic chips, reagents and consumables
Note: Incompatible with G2938A Bioanalyzer instruments
Catalog # 76645-152
Supplier #:  5067-4626
Bioanalyzer High Sensitivity DNA Analysis
Catalog # 76645-152
Bioanalyzer High Sensitivity DNA Analysis
Bioanalyzer High Sensitivity DNA Analysis
Agilent High sensitivity DNA kit, For the separation, sizing and quantification of dsDNA samples of limited abundance and sized from 50 - 7000 bp, the kit includes 10 microfluidic chips, reagents and consumables
Note: Incompatible with G2938A Bioanalyzer instruments
Price
$983.00
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Quality control of low input NGS libraries or DNA fragments up to 7000 bp.
  • Increased sensitivity for DNA analysis, down to 5 pg/µl for fragment analysis or 100 pg/µl for complex DNA samples, such as NGS libraries
  • A broad linear dynamic range that allows detection of less abundant PCR products
  • Accurate smear analysis within a specified size range of interest
  • Enhanced sizing accuracy, normalized to internal markers and external ladder
  • Quantitation accuracy and reproducibility calculated against precise internal standards
  • Minimal sample consumption is required as only 1 µl of sample is needed for analysis
  • Minimal exposure to hazardous materials, such as ethidium bromide, ensures safety
  • Achieve improved quality control analysis of next-generation sequencing (NGS) library smears derived from just a few amplification cycles
High Sensitivity DNA electrophoresis with the Bioanalyzer system enables improved quality control analysis of next-generation sequencing (NGS) library smears derived from just a few amplification cycles. Whether fragmented DNA or DNA sequencing libraries, the Bioanalyzer High Sensitivity DNA Kit reliably sizes and quantifies limited samples in the lower pg/µl concentration range.
The High Sensitivity DNA assays are often used for sample quality control for next-generation sequencing libraries. In addition, PCR amplicon analysis, multiplex PCR amplicons, gene expression analysis by RT-PCR, restriction fragment analysis, and detection of targeted cleavage in gene editing studies may all be carried out. Accuracy is ensured with a broad linear dynamic range and normalization to internal markers and ladder.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Documents
Specifications
  • Description:
    Agilent High sensitivity DNA kit
  • Details:
    For the separation, sizing and quantification of dsDNA samples of limited abundance and sized from 50 - 7000 bp, the kit includes 10 microfluidic chips, reagents and consumables
    Note: Incompatible with G2938A Bioanalyzer instruments