PRIMARY CELL PHENOL RED PH INDICATOR 1ML
Catalog # MSPP-PCS-999-0
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
PRIMARY CELL PHENOL RED PH INDICATOR 1ML
Catalog # MSPP-PCS-999-0
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: PCS-999-001
Frequently Bought Together
Vi-CELL BLU Reagent Kits, Beckman Coulter
Supplier: Beckman Coulter
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
DNA/RNA Shield™ Reagent, Zymo Research
Supplier: Zymo Research
Disposable PD-10 Desalting Columns, Cytiva
Supplier: Cytiva
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Acro® 50 Vent Filters, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
SOLUTION TRYPSIN NEUTRALIZING
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
CELL PRIMARY EPIDERMAL KERATINOCYTES 1ML
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Chemi Blot Blocking Buffer, Azure
Supplier: Azure Biosystems
KIT ENDOTHELIAL CELL GROWTH VEGF
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
RECOM FAS LIGAND/TNFSF6 HU >95% 10UG
Supplier: Novus Biologicals
KIT KERATINOCYTE GROWTH 0.06 MM FROZEN
Supplier: AMERICAN TYPE CULTURE COLLECTION MS