CHLORAC BUFFER LIQUID COLORLESS 250ML
Catalog # MSPP-1700-0132
Supplier: Pickering Labs
CHLORAC BUFFER LIQUID COLORLESS 250ML
Catalog # MSPP-1700-0132
Supplier: Pickering Labs
Supplier Number: 1700-0132
Frequently Bought Together
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
Dulbecco's phosphate-buffered saline (DPBS), Mediatech
Supplier: Corning
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Weighing Paper
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Sterile 70% Isopropanol
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® Disposable Petri Dishes
Supplier: VWR International
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International