Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT HUMAN PRL-R BIOTIN 200UG
Catalog # MSPP-23030076B
Supplier:
RAYBIOTECH LIFE INC MS
Expand image
RECOMBINANT HUMAN PRL-R BIOTIN 200UG
Catalog # MSPP-23030076B
Supplier:
RAYBIOTECH LIFE INC MS
Supplier Number:
230-30076B-200