Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ASSAY PLATE BLOCK 96 WELL FLAT BOTTOM
Catalog # MSPP-620-5020
Supplier:
TORREY PINES SCIENTIFIC MS
Expand image
ASSAY PLATE BLOCK 96 WELL FLAT BOTTOM
Catalog # MSPP-620-5020
Supplier:
TORREY PINES SCIENTIFIC MS
Supplier Number:
620-5020