CELL LINE EPITHELIAL MOUSE EO771 FROZEN
Catalog # MSPP-CRL-3461
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
CELL LINE EPITHELIAL MOUSE EO771 FROZEN
Catalog # MSPP-CRL-3461
Supplier: AMERICAN TYPE CULTURE COLLECTION MS
Supplier Number: CRL-3461
Frequently Bought Together
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Supplier: VWR
VWR® Absorbent Underpads with Waterproof Moisture Barrier
Supplier: VWR International
Disposable PD-10 Desalting Columns, Cytiva
Supplier: Cytiva
Mix & Go!™ E. coli Transformation Kit and Buffer Set
Supplier: Zymo Research
Amersham ECL Western Blotting Select Detection Reagent, Cytiva
Supplier: Cytiva
Western blotting detection reagent, Amersham™ ECL™ Prime
Supplier: Cytiva
Tris HCl pH 8.5, 1.0 M
Supplier: Rockland Immunochemical
DNA Wash Buffer, Concentrate, Zymo Research
Supplier: Zymo Research
BUFFER TBS BLOCKING READY-TO-USE 500ML
Supplier: Li-Cor
Radiance Plus Chemiluminescent Substrate, Azure Biosystems
Supplier: Azure Biosystems
Proteomelab™ SDS-MW Analysis Kit, SCIEX
Supplier: AB SCIEX LLC MS
Laemmli SDS sample buffer, reducing (6X), Thermo Scientific
Supplier: Thermo Scientific Chemicals
Dispase (1 U/ml)
Supplier: Stemcell Technologies
FlashGel™ DNA, RNA and Recovery Systems
Supplier: Lonza
Bis-Tris Mini Protein Gels, GenScript®
Supplier: Genscript
DNA BINDING BUFFER 100 ML
Supplier: Zymo Research