Orders LinkContactUsLinkComponent
Cubelets Accessories
Cubelets Accessories
Cubelets 5-port USB charger
Catalog # 470348-190
Supplier:   MODULAR ROBOTICS
Supplier #:  5PORTCHARGER
Cubelets Accessories
Catalog # 470348-190
Supplier:  MODULAR ROBOTICS
Cubelets Accessories
Cubelets Accessories
Cubelets 5-port USB charger
Price
$45.39
Log in for availability
Each
Product Details & Documents
Complete your Cubelets robotic system with these accessories.
  • Program your Cubelets using the Bluetooth Hat
  • Integrate your Cubelets with LEGO® bricks
  • Charge multiple Cubelets batteries with the USB charger
  • Contain and store your Cubelets with an attractive and rugged plastic bin
The Cubelets Buetooth Hat provides a connection between your Cubelets robot constructions and free Cubelets software. Use the Cubelets App to create remote control constructions, or Cubelets Console to visualize data, swap Personalities, and code new Personalities for your Cubelets. Program your Cubelets using Blockly or C coding.
Use the brick adapters to connect Cubelets and LEGO® bricks. The 4-pack includes two “studs” adapters and two “sockets” adapters. Offset placement allows for brick-connection across two adapters.
Charge up to five Cubelet batteries with the 5-port USB wall charger. Charges devices with 5V USB power input. Compact and portable for home or classroom use. Micro USB cables available separately.
The durable plastic carrying cases come in three colors, and include custom foam inserts to fit up to 24 Cubelets and accessories.
Specifications
  • Description:
    Cubelets 5-port USB charger