NEEDLE INSERT VELOS 34G MTL LOW FLOW KIT
Catalog # MSPP-OPTON-530
Supplier: THERMO ELECTRON NORTH AMERICA MS
NEEDLE INSERT VELOS 34G MTL LOW FLOW KIT
Catalog # MSPP-OPTON-530
Supplier: THERMO ELECTRON NORTH AMERICA MS
Supplier Number: OPTON-53011
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
pH Test Strips, VWR Chemicals BDH®
Supplier: BDH
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Pasteur Pipettes, Glass
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
Falcon® Centrifuge Tubes, Conical Bottom, Corning
Supplier: Corning
Tissue-Tek® O.C.T. Compound, Sakura® Finetek
Supplier: Sakura Finetek
VWR® Weighing Paper
Supplier: VWR International
Pierce™ Streptavidin Magnetic Beads, Thermo Scientific
Supplier: Invitrogen