Orders LinkContactUsLinkComponent
VWR® Cryo Coder Cap Inserts
VWR® Cryo Coder Cap Inserts
Cap insert, Yellow
Catalog # VWRI210-000039
Supplier:   VWR International
VWR® Cryo Coder Cap Inserts
Catalog # VWRI210-000039
Supplier:  VWR International
VWR® Cryo Coder Cap Inserts
VWR® Cryo Coder Cap Inserts
Cap insert, Yellow
Price
$48.56
Log in for availability
Case of 500
Product Details & Documents
Product can be used as coding/identification of samples types stored in it.
Cryocoder/cap inserts are available in multiple color options: Violet, white, blue, orange, lilac, red, tan, yellow, green, grey, pink, assorted (five colours).
Specifications
  • Color:
    Yellow
  • Description:
    Cap insert