Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SENSOR TEMP 230MM GLASS F GHS HEAT MODEL
Catalog # 76595-400
Supplier:
Globe Scientific
Expand image
SENSOR TEMP 230MM GLASS F GHS HEAT MODEL
Catalog # 76595-400
Supplier:
Globe Scientific
Supplier Number:
GHS-S230G