Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Heated Forceps System, Mopec

Heated Forceps System, Mopec

Heated forceps system, Smooth
Catalog # 76579-224
Supplier:   Mopec
Supplier #:  BK300-SM
Heated Forceps System, Mopec
Catalog # 76579-224
Supplier:  Mopec
Heated Forceps System, Mopec
Heated Forceps System, Mopec

Heated forceps system, Smooth

Price
$2,678.33
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Mopec’s heated forceps system is designed to take up minimal bench-top space and comes complete with a power supply, control unit, and two different heated forceps. The ergonomic design ensures user comfort while performing specimen placement, and the anti-microbial coating helps prevent bacteria and virus growth.

  • Choose either serrated or smooth tips
  • Designed to take up minimal bench-top space
  • Anti-microbial coating to help prevent bacteria and virus growth

Sold individually.

Delivery information: System includes:
1 mm Heated forceps (Red)
2 mm Heated forceps (Yellow)
BK330: Mopec control unit
BK320: Mopec power supply

Specifications
  • Description:
    Heated forceps system
  • Tip style:
    Smooth

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.