Large Pathology Containers, Mopec
Product Details & Documents
Mopec’s screw top pathology containers are made from a high-quality, clarified polypropylene material. The screw-top lid creates a moisture and tamper-resistant closure that increases safety for laboratory employees and helps protect pathology specimens during transportation. Features an attached handle for easy transportation. Available in multiple sizes.
- Made from a high-quality, clarified polypropylene material
- Screw-top lid creates a moisture and tamper-resistant closure
- Features an attached handle for easy transportation
Sold individually.
Specifications
- Capacity:1 gal.
- Description:Large pathology containers