Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

Glass bottle for solids, 5 g
Catalog # TS22928-0050
Irritant
Health
Supplier #:  22928-0050
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
Catalog # TS22928-0050
CAS Number:  54827-17-7
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%
3,3',5,5'-Tetramethylbenzidine (TMB) 99+%

Glass bottle for solids, 5 g

CAS Number:  54827-17-7
Price
$175.00
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
  • CAS Number:54827-17-7
  • Formula:C₁₆H₂₀N₂
  • MDL Number:MFCD00007748
  • Molecular Weight:240.35 g/mol
  • Melting Pt:167…170 °C
  • Storage Temperature:Ambient
  • EINECS:259-364-6
Specifications

Specification Test Results

  • Appearance:
    White to cream to light yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    167 to 172 °C
  • Loss on drying:
    ≤0.5 %
  • Sulfated ash:
    ≤0.2 %
  • HPLC:
    ≥99.0 %
  • Titration with HClO4:
    98.0 to 102.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.