Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STORAGE BOX PP FOR 4ML VIAL 130X130X52MM
Catalog # MSPP-AAV263235
Supplier:
ANALYTICS-SHOP USA LP MS
Expand image
STORAGE BOX PP FOR 4ML VIAL 130X130X52MM
Catalog # MSPP-AAV263235
Supplier:
ANALYTICS-SHOP USA LP MS
Supplier Number:
AAV26323537