$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
ALNICO PR 1.5 W/KEEPER IND 2PK
470346-026
:
Wards
ALNICO PR 1.5 W/KEEPER IND 2PK
470346-026
:
Wards
:
PH0789N8