Orders LinkContactUsLinkComponent
Eisco High-Performance Variable Volume Micropipettes
Eisco High-Performance Variable Volume Micropipettes
20 - 200 µl, 1.0 µl, Yellow, 1
Catalog # 470345-366
Supplier:   Wards
Supplier #:  VVM9
Eisco High-Performance Variable Volume Micropipettes
Catalog # 470345-366
Supplier:  Wards
Eisco High-Performance Variable Volume Micropipettes
Eisco High-Performance Variable Volume Micropipettes
20 - 200 µl, 1.0 µl, Yellow, 1
Price
$172.85
Log in for availability
Each
Product Details & Documents
Variable volume micropipettes are fully autoclavable and feature a simple ergonomic design for comfort, color coded plungers, and calibration conforming to DIN ISO standards.
  • Soft and lightweight operation with an ergonomic, soft touch tip ejector designed to minimize hand fatigue, reduce thumb stress and to increase comfort with prolonged use
  • Fully autoclavable with a durable, autoclavable PVDF tip cone
  • Slip-resistant thermoplastic elastomer (TPE) body grip to minimize body heat transfer to the micropipette during long use, resulting in higher accuracy
  • Color coded plungers for easy identification
  • Calibration conforms to DIN 12650 & EN-ISO 8655 standard ensuring high accuracy and precision - Calibration report enclosed with every micropipette
High performance variable volume micropipettes have a simple design which allows for easy adjustment and reading of increments.
Specifications
  • Volume:
    20 - 200 µl
  • Increments:
    1.0 µl
  • No. of channels:
    1
  • Color code:
    Yellow