$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
TUNING FORK SET - ALUMINIUM
470344-556
:
Wards
TUNING FORK SET - ALUMINIUM
470344-556
:
Wards
:
PH0744B