Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
BAR MAGNETS W KEEPERS 2/PK
470343-966
:
Wards
BAR MAGNETS W KEEPERS 2/PK
470343-966
:
Wards
:
PH0786D