Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CONCAVE MIRROR - GLASS 38MM
470343-938
:
Wards
CONCAVE MIRROR - GLASS 38MM
470343-938
:
Wards
:
PH0519B