$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
CHROME STEEL HORSESHOE MAGNET
470343-792
:
Wards
CHROME STEEL HORSESHOE MAGNET
470343-792
:
Wards
:
PH0780C