Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Eisco Plastic Filter Funnels

Eisco Plastic Filter Funnels

1, 150 mm, 110 mm
Catalog # 470343-784
Supplier:   Wards
Supplier #:  CH0454E
Eisco Plastic Filter Funnels
Catalog # 470343-784
Supplier:  Wards
Eisco Plastic Filter Funnels
Eisco Plastic Filter Funnels

1, 150 mm, 110 mm

Price
$3.85
Log in for availability
Each
Product Details & Documents

Chemically-resistant filter funnels made of high quality polypropylene plastic.

  • Excellent resistance against acids, alcohols, aldehydes, bases, and hydrocarbons
  • Useful for separating solids from liquids, as well as for easier transfers of liquids to narrow mouthed containers
  • Autoclavable and microwavable

Packaging: Pack of 1 or 10.

Specifications
  • Ø:
    150 mm
  • Size:
    1
  • Stem length:
    110 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.