Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
InVivoPure™ pH 8.0 Dilution Buffer, BioXCell

InVivoPure™ pH 8.0 Dilution Buffer, BioXCell

1X, 50 ml, 8.0
Catalog # 76555-584
Supplier:   Bio X Cell
Supplier #:  IP0080-50ML-
InVivoPure™ pH 8.0 Dilution Buffer, BioXCell
Catalog # 76555-584
Supplier:  Bio X Cell
CAS Number:  
InVivoPure™ pH 8.0 Dilution Buffer, BioXCell

1X, 50 ml, 8.0

CAS Number:  
Price
$109.88
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

InVivoPure™ dilution buffers are specifically formulated and tested to satisfy the stringent requirements for in vivo applications.

They are extremely low in endotoxin, have been screened for murine pathogens, tested in animal models for toxicity and are formulated with respect to buffer composition and pH to satisfy the requirements of Bio X Cell's antibodies.

This buffer does not contain calcium, magnesium, phenol red, or preservatives such as azide.

Caution: Keep contents sterile. Open only in a biological safety cabinet.

Specifications

Specifications

  • Concentration:
    1X
  • pH:
    8.0
  • Volume:
    50 ml
  • Cat. No.:
    76555-584
  • Shelf Life:
    One year from the date of customer receipt when stored as recommended.
  • Sterility:
    0.2 µM filtered
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.