Mouse IgG Isotype Control
100 mg
Catalog # 76554-526
Supplier: Bio X Cell
Supplier #: BE0093-100MG
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
InVivoMab polyclonal mouse IgG, Clone: Polyclonal, Isotype: mouse IgG, Applications: in vivo administration
- Recommended Dilution Buffer: IP0070
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Specifications
- Size:100 mg
- Cat. No.:76554-526
- Clonality:Polyclonal
- Antibody Type:Primary
- Isotype:IgG
- Host:Mouse
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



