Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
ω-Bromo-3-nitroacetophenone 97%

ω-Bromo-3-nitroacetophenone 97%

Glass bottle for solids, 25 g
Catalog # TS13747-0250
Corrosive
Supplier #:  13747-0250
ω-Bromo-3-nitroacetophenone 97%
Catalog # TS13747-0250
CAS Number:  2227-64-7
ω-Bromo-3-nitroacetophenone 97%
ω-Bromo-3-nitroacetophenone 97%

Glass bottle for solids, 25 g

CAS Number:  2227-64-7
Price
$73.90
Log in for availability
Each (25g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  3-Nitrophenacyl bromide, 2-Bromo-3'-nitroacetophenone, α-Bromo-3-nitroacetophenone, Bromomethyl m-nitrophenyl ketone
  • CAS Number:2227-64-7
  • Formula:C₈H₆BrNO₃
  • MDL Number:MFCD00024512
  • Molecular Weight:244.04 g/mol
  • Melting Pt:90…94 °C
  • Storage Temperature:Ambient
  • EINECS:218-764-0
  • UN:3261
  • ADR:8,II
Specifications

Specification Test Results

  • Appearance:
    Yellow-green to beige to light orange Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    90 to 94 °C
  • GC:
    ≥96.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    25 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.