Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
2'-Methylacetophenone 98%

2'-Methylacetophenone 98%

Glass bottle, 5 g
Catalog # TS12616-0050
Supplier #:  12616-0050
2'-Methylacetophenone 98%
Catalog # TS12616-0050
CAS Number:  577-16-2
2'-Methylacetophenone 98%
2'-Methylacetophenone 98%

Glass bottle, 5 g

CAS Number:  577-16-2
Price
$33.00
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  o-Methylacetophenone, Methyl o-tolyl ketone, 1-(o-Tolyl)ethanone
  • CAS Number:577-16-2
  • Formula:C₉H₁₀O
  • MDL Number:MFCD00008734
  • Molecular Weight:134.18 g/mol
  • Boiling Pt:214 °C (760 mmHg)
  • Density:1.026 g/cm³
  • Flash Pt:75 °C
  • Storage Temperature:Ambient
  • EINECS:209-408-5
Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5290 to 1.5330 (20 °C, 589 nm)
  • GC:
    ≥97.5 %
  • UV transmittance:
    ≥99 % (c=0.01M water, 254 nm)

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.