Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Maltol 99%

Maltol 99%

Glass bottle for solids, 5 g
Catalog # TS12155-0050
Irritant
Supplier #:  12155-0050
Maltol 99%
Catalog # TS12155-0050
CAS Number:  118-71-8
Maltol 99%
Maltol 99%

Glass bottle for solids, 5 g

CAS Number:  118-71-8
Price
$23.10
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  3-Hydroxy-2-methyl-γ-pyrone, Maltol, 3-Hydroxy-2-methyl-4-pyranone, 3-Hydroxy-2-methyl-4H-pyran-4-one, 3-Hydroxy-2-methyl-4-pyrone
  • CAS Number:118-71-8
  • Formula:C₆H₆O₃
  • MDL Number:MFCD00006578
  • Molecular Weight:126.11 g/mol
  • Merck Index:13,05735
  • Boiling Pt:170 °C (100 mmHg)
  • Melting Pt:160…164 °C
  • Storage Temperature:Ambient
  • EINECS:204-271-8
Specifications

Specification Test Results

  • Appearance:
    White to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    160 to 164 °C
  • Water:
    ≤0.5 %
  • GC:
    ≥98.5 %
  • Arsenic (As):
    ≤3 ppm
  • Lead (Pb):
    ≤10 ppm
  • UV:
    at 220 nm A: ≤0.2
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤2 ppm
  • Acidity/alkalinity:
    ≤0.0001 meq/g

Specifications

  • Pack type:
    Glass bottle for solids
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.