Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RECOMBINANT CD71 TFRC HUMN PROTEIN 100UG
Catalog # MSPP-TP326147M
Supplier:
Origene Technologies
Expand image
RECOMBINANT CD71 TFRC HUMN PROTEIN 100UG
Catalog # MSPP-TP326147M
Supplier:
Origene Technologies
Supplier Number:
TP326147M