Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

L(+)-Ascorbic acid 99% ACS

Plastic bottle, 2.5 kg
Catalog # TS40147-0025
Irritant
Health
Flammable
 :  40147-0025
L(+)-Ascorbic acid 99% ACS
Catalog # TS40147-0025
CAS Number:  50-81-7
L(+)-Ascorbic acid 99% ACS
L(+)-Ascorbic acid 99% ACS

Plastic bottle, 2.5 kg

CAS Number:  50-81-7
Price
$1,200.00
Log in for availability
Each (2,500g/mol)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Ascorbic Acid, L-Ascorbic acid, (R)-5-((S)-1,2-Dihydroxyethyl)-3,4-dihydroxyfuran-2(5H)-one, Acidum ascorbicum
  • CAS Number:50-81-7
  • Formula:C₆H₈O₆
  • MDL Number:MFCD00064328
  • Molecular Weight:176.13 g/mol
  • Merck Index:13,00837
  • Boiling Pt:62 °C (1013 hPa)
  • Melting Pt:–136 °C (dec.)
  • Density:0.68 g/cm³
  • Flash Pt:–26 °C
  • Storage Temperature:Refrigerator
  • EINECS:212-108-7
  • UN:3295
  • ADR:3,II
  • REACH:exempt
Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3910 to 1.3930 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Specific optical rotation:
    +20.5° to +21.5° (25°C, 589 nm) (c=10, H2O)
  • Iron (Fe):
    ≤10 ppm
  • Heavy metals (ICP-OES):
    ≤20 ppm

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    2.5 kg

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.