Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

3-(4-Bromophenyl)propionic acid 97%

Glass bottle, 5 g
Catalog # TS39945-0050
Irritant
 :  39945-0050
3-(4-Bromophenyl)propionic acid 97%
Catalog # TS39945-0050
CAS Number:  1643-30-7
3-(4-Bromophenyl)propionic acid 97%
3-(4-Bromophenyl)propionic acid 97%

Glass bottle, 5 g

CAS Number:  1643-30-7
Price
$103.00
Log in for availability
Each (5g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
  • CAS Number:1643-30-7
  • Formula:C₉H₉BrO₂
  • MDL Number:MFCD01310793
  • Molecular Weight:229.07
  • Melting Pt:133…136 °C
  • Storage Temperature:Ambient
  • UN:3335
  • ADR:9,III
Specifications

Specification Test Results

  • Appearance:
    White to yellow to beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    134 to 138 °C
  • HPLC:
    ≥96.0 %

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 g

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.