Orders LinkContactUsLinkComponent
Masterflex® Liquid Detector Pad Cables, Avantor®
Liquid Detector Pad; 3.5 ft Cable
  MFLX77095-00
Masterflex® Liquid Detector Pad Cables, Avantor®
  MFLX77095-00
 :  
Masterflex® Liquid Detector Pad Cables, Avantor®
Masterflex® Liquid Detector Pad Cables, Avantor®
Liquid Detector Pad; 3.5 ft Cable
Make the most of your Liqui-Sense leak detector with suitable replacement parts.
  • Compatible with Masterflex® Liqui-Sense controllers
This replacement cable features a 6-pin cable with RJ-12 connections and has a 3.5 foot cable length.
  • Description:
    Liquid Detector Pad; 3.5 ft Cable
Accessories
  •  
    Description
    Length
  • MFLX77095-02
    Replacement RJ-12 cable
    3.5 ft.
    Case (3.5Feet) - $79.40