Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Hypodermic Needles, Mortech

Hypodermic Needles, Mortech

13, 114.3 mm (4.50")
Catalog # 76549-814
Supplier:   Mopec
Supplier #:  FC912-24
Hypodermic Needles, Mortech
Catalog # 76549-814
Supplier:  Mopec
Hypodermic Needles, Mortech
Hypodermic Needles, Mortech

13, 114.3 mm (4.50")

Price
$10.97
Log in for availability
Each
Product Details & Documents

Hypodermic needles are designed for aspiration/cavity injection or extraction.

  • Stainless steel, long lasting piercing points, Luer hubs.

Available in a variety of sizes.

Specifications
  • Length:
    114.3 mm (4.50")
  • Gauge:
    13
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.